EMAIL THIS PAGE TO A FRIEND
Nature chemistry
Polymer synthesis: facilitating functionality.
PMID 21124472
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210145
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
C6H8O4