EMAIL THIS PAGE TO A FRIEND
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases
[Investigation on coal work's production use of chloroacetic acid in one province].
PMID 21126433