EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Tripterygium wilfordii Hook F. versus Sulfasalazine in the treatment of rheumatoid arthritis: a well-designed clinical trial of a botanical demonstrating effectiveness.
PMID 21126560
Abstract
Claims that Traditional/Alternative medicine (TM/AM) remedies are effective are routinely ignored by Western Medicine. However, the results of a clinical trial that demonstrated the clinical efficacy of Tripterygium wilfordii Hook F. (TW), a TM used as an anti-inflammatory, were recently published in the Annals of Internal Medicine. Why this article was published in a peer reviewed Allopathic medical journal is an important question that begs examination and may lay the future promise of TM/AM therapeutic interventions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0883
Sulfasalazine, analytical standard, ≥98% (HPLC)
C18H14N4O5S