EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
MDCK cell permeability characteristics of a sulfenamide prodrug: strategic implications in considering sulfenamide prodrugs for oral delivery of NH-acids.
PMID 21126873
Abstract
The objective of this Letter is both to report the permeability results of a linezolid-based sulfenamide prodrug in an MDCK cell model (enterocyte surrogate system) and to discuss the strategic implications of these results for considering sulfenamide prodrugs to enhance the oral delivery of weakly acidic NH-acids (e.g., amides, ureas, etc.). The two main findings from this study are that the sulfenamide prodrug does not appear to survive intracellular transport due to conversion to linezolid and that there appears to be an apically-oriented surface conversion pathway that can additionally serve to convert the sulfenamide prodrug to linezolid upon approach of the apical membrane. It is hoped that these findings, along with the discussion of the strategic implications, will facilitate a greater awareness of the potential strengths and weaknesses inherent in the sulfenamide prodrug approach for enhancing the oral delivery of weakly acidic NH-acid drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8876
Sulfamerazine, ReagentPlus®, ≥99.0%
C11H12N4O2S
46826
Sulfamerazine, VETRANAL, analytical standard
C11H12N4O2S