EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Semisynthesis of 6-chloropurine-2'-deoxyriboside 5'-dimethoxytrityl 3'-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidite and its use in the synthesis of fluorescently labeled oligonucleotides.
PMID 21128170
Abstract
An efficient enzymatic synthesis of 6-chloropurine-2'-deoxyriboside from the reaction of 6-chloropurine with 2'-deoxycytidine catalyzed by nucleoside-2'-deoxyribosyltransferase (E.C. 2.4.2.6) followed by chemical conversion into the 5'-dimethoxytrityl 3'-(2-cyanoethyl-N,N-diisopropylamino) phosphoramidite derivative is described. The phosphoramidite derivative was incorporated site-specifically into an oligonucleotide and used for the introduction of a tethered tetramethylrhodamine-cadaverine conjugate. The availability of an efficient route to 6-chloropurine-2'-deoxyriboside 5'-dimethoxytrityl 3'-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidite enables the facile synthesis of oligonucleotides containing a range of functional groups tethered to deoxyadenosine residues.