EMAIL THIS PAGE TO A FRIEND
Free radical research
Synthesis and antioxidant capacities of hydroxyl derivatives of cinnamoylphenethylamine in protecting DNA and scavenging radicals.
PMID 21128731
Abstract
Cinnamoylphenethylamine (CNPA) derivatives including feruloylphenethylamine (FRPA), caffeoylphenethylamine (CFPA), cinnamoyltyramine (CNTA), feruloyltyramine (FRTA) and caffeoyltyramine (CFTA) were synthesized in order to investigate the influence of the number and position of hydroxyl group on Cu(2+)/glutathione (GSH) and 2,2'-azobis(2-amidinopropane hydrochloride) (AAPH)-induced oxidation of DNA. The radical-scavenging properties of these CNPA derivatives were also evaluated by trapping 2,2'-azinobis(3-ethylbenzothiazoline-6-sulphonate) cationic radical (ABTS(+•)), 2,2'-diphenyl-1-picrylhydrazyl radical (DPPH) and galvinoxyl radical. In addition, these CNPA derivatives were tested by linoleic acid (LH)-β-carotene-bleaching experiment. The chemical kinetic was employed to treat the results from AAPH-induced oxidation of DNA and gave the order of antioxidant ability as CFTA > CFPA > FRTA > FRPA. CFTA and CFPA also possessed high abilities to inhibit Cu²(+)/GSH-mediated degradation of DNA, whereas FRPA and FRTA can protect LH against the auto-oxidation efficiently. Finally, CFPA and FRPA exhibited high activity in trapping ABTS(+•), DPPH and galvinoxyl radicals. Therefore, the cinnamoyl group bearing ortho-dihydroxyl or hydroxyl with ortho-methoxyl benefited for CNPA derivatives to protect DNA, while hydroxyl in tyramine cannot enhance the radical-scavenging abilities of CNPA derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G307
Galvinoxyl, free radical
C29H41O2