EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Proniosomes as a carrier system for transdermal delivery of tenoxicam.
PMID 21129461
Abstract
Tenoxicam is a non steroidal anti-inflammatory drug (NSAID) widely used in the treatment of rheumatic diseases and characterized by its good efficacy and less side effects compared to other NSAIDs. Its oral administration is associated with severe side effects in the gastrointestinal tract. Transdermal drug delivery has been recognized as an alternative route to oral delivery. Proniosomes offer a versatile vesicle delivery concept with the potential for drug delivery via the transdermal route. In this study, different proniosomal gel bases were prepared, characterized by light microscopy, revealing vesicular structures, and assessed for their drug entrapment efficiency, stability, their effect on in vitro drug release and ex vivo drug permeation. The lecithin-free proniosomes prepared from Tween 20:cholesterol (9:1) proved to be stable with high entrapment and release efficiencies. The in vivo behaviour of this formula was studied on male rats and compared to that of the oral market product. The investigated tenoxicam loaded proniosomal formula proved to be non-irritant, with significantly higher anti-inflammatory and analgesic effects compared to that of the oral market tenoxicam tablets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0909
Tenoxicam, NSAID
C13H11N3O4S2
T0040800
Tenoxicam, European Pharmacopoeia (EP) Reference Standard
C13H11N3O4S2