EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Development of a polyoxometallate-based photocatalyst assembled with cucurbit[6]uril via hydrogen bonds for azo dyes degradation.
PMID 21130567
Abstract
A water insoluble cucurbit[6]uril-polyoxometallates (CB[6]-POMs) composite assembled from α-Keggin type polysilicontungstate anions and macrocycle cucurbit[6]uril (CB[6]) via hydrogen bonding has been synthesized as visible light active photocatalyst. The physical and photocatalytic properties of such photocatalyst have been fully characterized by PXRD, FTIR, TG, XPS, and UV/vis diffuse reflectance spectra. The catalyst shows a good photocatalytic activity towards the degradation of methyl orange (MO) under visible light irradiation and displays good reproducibility of photocatalytic degradation by a simple recycled procedure without obvious loss in catalytic activity, which is of great significance for practical use of the photocatalyst. In the photodegradation process, the {Ni-CB[6]}(n) chain of the photocatalyst acts as sensitizer and can be induced by visible light, meanwhile the POMs chain of the photocatalyst acts as electron acceptor and deposits the electron in its LUMO. The effects of various experimental parameters and the proposed mechanisms are discussed in detail.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94544
Cucurbit[6]uril hydrate, contains acid of crystalization
C36H36N24O12 · xH2O