EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of 7-substituted-1-(3-bromophenylamino)isoquinoline-4-carbonitriles as inhibitors of myosin light chain kinase and epidermal growth factor receptor.
PMID 21130659
Abstract
Here we present the synthesis and biological activity of a series of 7-substituted-1-(3-bromophenylamino)isoquinoline-4-carbonitriles as inhibitors of myosin light chain kinase (MLCK) and the epidermal growth factor receptor kinase (EGFR). The inhibitory effect of these molecules was found to be dependent on the nature of the substituents at the 7-position of the isoquinoline scaffold.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K2015
K-252a, Ready Made Solution, from Nonomuraea longicatena, >98%
C27H21N3O5