EMAIL THIS PAGE TO A FRIEND
Metallomics : integrated biometal science
Synthesis, structure and biological evaluation of bis salicylaldehyde-4(N)-ethylthiosemicarbazone ruthenium(iii) triphenylphosphine.
PMID 21132183
Abstract
Bis salicylaldehyde-4(N)-ethylthiosemicarbazone ruthenium(iii) triphenylphosphine [Ru(Sal-etsc)(H-Sal-etsc)(PPh(3))] was synthesized and structurally characterized by spectral and X-ray crystallographic studies and it showed 100% inhibition on the DPPH radical. It also exhibited a significant lymphocyte activity and inhibitory effect on the lung carcinoma A549 cell.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
658774
Salicylaldehyde thiosemicarbazone, 95%
C8H9N3OS