EMAIL THIS PAGE TO A FRIEND
Acta medica Iranica
Traffic noise exposure increases gastric acid secretion in rat.
PMID 21132997
Abstract
Noise is considered as one of the most severe sources of environmental and work place constraints. Noise effects on immune function, hormonal levels, cardiovascular and respiratory systems are well known. The aim of the study is to evaluate the effects of traffic noise on basal and stimulated gastric acid secretion. 48 healthy rats were divided into five traffic noise exposures (1, 7, 14, 21, 28 days) and a control groups. Pentagastrin was used i.p. for stimulation of gastric acid secretion. The gastric contents were collected by the wash-out technique and then titrated. Histological studies were performed on gastric epithelial layer. In the 1, 7, 14 and 21 days traffic noise exposure, basal and pentagastrin-stimulated gastric acid secretion increased compared to the control group (P<0.001), but a significant decrease was seen in hyperacidity in 28th days, in the both basal and stimulated states (P<0.05). Histological study showed that mucosal layer thickness of stomach increased, while the number of oxyntic glands and cell nuclei decreased. It seems that 1,7,14 and 21 days traffic noise increase gastric acid secretion, while 28 days traffic noise can induce adaptation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1636
Pentagastrin, ≥95% (HPLC), powder
C37H49N7O9S