EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Precipitation of dispersed silver particles using acetone as reducing agent.
PMID 21134680
Abstract
A novel environmentally friendly solution-based method for preparing dispersed silver particles is described. The simple and convenient approach consists in heating silver oxide particles dispersed in a highly alkaline water/acetone mixture. The data presented clearly show that acetone reduces completely and rapidly Ag(2)O particles to metallic silver at 60°C. A mechanism explaining the provenance of the electrons responsible for the reduction of silver is proposed.