EMAIL THIS PAGE TO A FRIEND
Chemosphere
The dissipation rates of trichlorfon and its degradation product dichlorvos in cabbage and soil.
PMID 21134689
Abstract
The residual levels and dissipation rate of trichlorfon, and its degradation product, dichlorvos, in cabbage crops and the soil in which these were grown, were determined by gas chromatography at two geographically distant experimental sites, one in Kunming and one in Beijing, China. Trichlorfon was applied at two dosages (900 g ai ha(-1) and 1350 g ai ha(-1)). Maximum final residues of trichlorfon in soil and cabbage were 1.23 mg kg(-1) and 1.81 mg kg(-1) respectively at Kunming, and 0.35 mg kg(-1) and 0.70 mg kg(-1) respectively at Beijing. However, the final residues of dichlorvos in both cabbage and soil was only 0.04 mg kg(-1) at Kunming, and only 0.03 mg kg(-1), or "not detectable", at Beijing. The mean half-life of trichlorfon in cabbage was 1.80 d with a dissipation rate of 90% over 5 d, while that in soil was 3.05 d with a dissipation rate of 90% over 14 d at one experimental site. The dissipation rates of trichlorfon and its degradation product dichlorvos at the two experimental sites were different, suggesting that degradation of these pesticides was affected by local soil characteristics and climate. When applied at both the recommended dosage and at 1.5 times this, no detectable residues of either trichlorfon or dichlorvos were found in soil or cabbage at harvest. Although trichlorfon can easily degrade into dichlorvos, which is highly toxic to humans and other animals, the observed low residual levels of dichlorvos suggest that trichlorfon is safe when applied at the recommended dosage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45698
Trichlorfon, PESTANAL®, analytical standard
C4H8Cl3O4P