EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents of radix Boehmeriae].
PMID 21137364
Abstract
To study the chemical constituents from Radix Boehmeriae. Compounds were isolated by various column chromatographies with silica gel, their structures were elucidated by spectral analysis and chemical evidence. Eight compounds were isolated and their structures were identified as emodin (1), emodin-8-O-beta-glucopyranoside (2), physcion (3), polydatin (4), catechin (5), epicatechin (6), potassium nitrate (7), sitosterol (8). Among these compounds, 3 -7 are isolated from Radix Boehmeriae for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17797
Physcion, ≥98.0% (TLC)
C16H12O5
93893
Physcion, analytical standard
C16H12O5