EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Orphan drugs. Regulation is flawed.
PMID 21139004
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7148
3,4-Diaminopyridine, ≥98%
C5H7N3