EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
New isocourmarin and phthalide derivatives from the rhizomes of Matteuccia orientalis.
PMID 21139273
Abstract
Five new compounds (1-5), were isolated from the rhizomes of Matteuccia orientalis (HOOK.) TREV. The structures of new compounds were elucidated on the basis of their 1D-, 2D-NMR, MS, IR and circular dichroism (CD) data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P39605
Phthalide, 98%
C8H6O2
W419501
Phthalide, 98%
C8H6O2