EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Treatment of hyperuricemia and gout based on the guideline for the management of hyperuricemia and gout].
PMID 21139283