EMAIL THIS PAGE TO A FRIEND
Journal of environmental monitoring : JEM
Presence and partitioning properties of the flame retardants pentabromotoluene, pentabromoethylbenzene and hexabromobenzene near suspected source zones in Norway.
PMID 21140013
Abstract
The brominated flame retardants (BFRs), pentabromotoluene (PBT), pentabromoethylbenzene (PBEB) and hexabromobenzene (HBB), exhibit physical-chemical properties similar to other persistent organic pollutants, and have been in use as flame retardants for several decades. Data on these BFRs in diverse environmental samples can be found in studies from the 1970s and 1980s, as well as in recent years, though very little in the years in between. Due to a lack of data, the cause for the apparent re-emergence of these BFRs in recent studies is unclear, and could reflect changes in production volumes, accumulation of transformation products from BFR precursors, improved analytical techniques or simply a re-emergence in concern. Very little data are available on their environmentally relevant partitioning properties, which could help to explain the occurrence and fate of these BFRs. In this study we analysed for the presence of HBB, PBT, and PBEB in diverse environmental samples from potential Norwegian BFR source zones. Additionally, environmental partitioning properties of these BFRs as well as brominated benzenes were estimated and validated using experimental data for brominated benzenes. Of the three BFRs, HBB was identified in detectable quantities at most source zones, PBEB only near a metal recycling factory, and PBT only in a few additional locations from where PBEB was detected. Data from this study show that HBB is likely widely distributed, as verified both by chemical analysis and estimated properties. Measured HBB levels in wastewater treatment plants indicate that the treatment practices used in the study locations are not effective at lowering HBB levels, perhaps due to association with low density suspended solids (e.g. microplastics).