EMAIL THIS PAGE TO A FRIEND
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Copper depletion increases the mitochondrial-associated SOD1 in neuronal cells.
PMID 21140194
Abstract
The role of copper in the toxicity of mutant copper-dependent enzyme superoxide dismutase (SOD1) found in patients affected with the familial form of amyotrophic lateral sclerosis (fALS) is widely debated. Here we report that treatment of human neuroblastoma cells SH-SY5Y with a specific copper chelator, triethylene tetramine (Trien) induces the decrease of intracellular copper level, paralleled by decreased activity of SOD1. A comparable effect is observed in mouse NSC-34-derived cells, a motoneuronal model, transfected for the inducible expression of either wild-type or G93A mutant human SOD1, one of the mutations associated with fALS. In both cell types, the drop of SOD1 activity is not paralleled by the same extent of decrease in SOD1 protein content. This discrepancy can be explained by the occurrence of a fraction of copper-free SOD1 upon copper depletion, which is demonstrated by the partial recovery of the enzyme activity after the addition of copper sulphate to homogenates of SH-SY5Y cells. Furthermore, copper depletion produces the enrichment of the physiological mitochondrial fraction of SOD1 protein, in both cells models. However, increasing the fraction of mitochondrial, possibly copper-free, mutant human SOD1 does not further alter mitochondrial morphology in NSC-34-derived cells. Thus, copper deficiency is not a factor which may worsen mitochondrial damage, which is one of the earliest events in fALS associated with mutant SOD1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90460
Triethylenetetramine, ≥97.0% (T)
C6H18N4
132098
Triethylenetetramine, technical grade, 60%
C6H18N4