EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Multiple fixed drug eruption caused by ornidazole.
PMID 21144346
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O5879
Ornidazole, analytical standard
C7H10ClN3O3