EMAIL THIS PAGE TO A FRIEND
Medicina clinica
[Urea as therapeutic option for the treatment of the syndrome of inappropriate antidiuretic hormone secretion].
PMID 21145071
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6140
Demeclocycline hydrochloride, powder or crystals
C21H21ClN2O8 · HCl
46161
Demeclocycline hydrochloride hydrate, VETRANAL®, analytical standard
C21H21ClN2O8 · HCl · xH2O