EMAIL THIS PAGE TO A FRIEND
Brain structure & function
Geometric and functional architecture of visceral sensory microcircuitry.
PMID 21153904
Abstract
Is microcircuit wiring designed deterministically or probabilistically? Does geometric architecture predict functional dynamics of a given neuronal microcircuit? These questions were addressed in the visceral sensory microcircuit of the caudal nucleus of the tractus solitarius (NTS), which is generally thought to be homogeneous rather than laminar in cytoarchitecture. Using in situ hybridization histochemistry and whole-cell patch clamp recordings followed by neuronal reconstruction with biocytin filling, anatomical and functional organization of NTS microcircuitry was quantified to determine associative relationships. Morphologic and chemical features of NTS neurons displayed different patterns of process arborization and sub-nuclear localization according to neuronal types: smaller cells featured presynaptic local axons and GABAergic cells were aggregated specifically within the ventral NTS. The results suggested both a laminar organization and a spatial heterogeneity of NTS microcircuit connectivity. Geometric analysis of pre- and postsynaptic axodendritic arbor overlap of reconstructed neurons (according to parent somal distance) confirmed a heterogeneity of microcircuit connectivity that could underlie differential functional dynamics along the dorsoventral axis. Functional dynamics in terms of spontaneous and evoked postsynaptic current patterns behaved in a strongly location-specific manner according to the geometric dimension, suggesting a spatial laminar segregation of neuronal populations: a dorsal group of high excitation and a ventral group of balanced excitation and inhibition. Recurrent polysynaptic activity was also noted in a subpopulation of the ventral group. Such geometric and functional laminar organization seems to provide the NTS microcircuit with both reverberation capability and a differentiated projection system for appropriate computation of visceral sensory information.