EMAIL THIS PAGE TO A FRIEND
Alternative medicine review : a journal of clinical therapeutic
Pantethine. Monograph.
PMID 21155629
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2125
D-Pantethine, analytical standard
C22H42N4O8S2