EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
ChIP-on-chip analysis reveals angiopoietin 2 (Ang2, ANGPT2) as a novel target of steroidogenic factor-1 (SF-1, NR5A1) in the human adrenal gland.
PMID 21163858
Abstract
The nuclear receptor steroidogenic factor-1 (SF-1, NR5A1) is a key regulator of adrenal and gonadal biology. Disruption of SF-1 can lead to disorders of adrenal development, while increased SF-1 dosage has been associated with adrenocortical tumorigenesis. We aimed to identify a novel subset of SF-1 target genes in the adrenal by using chromatin immunoprecipitation (ChIP) microarrays (ChIP-on-chip) combined with systems analysis. SF-1 ChIP-on-chip was performed in NCI-H295R human adrenocortical cells using promoter tiling arrays, leading to the identification of 445 gene loci where SF-1-binding regions were located from 10 kb upstream to 3 kb downstream of a transcriptional start. Network analysis of genes identified as putative SF-1 targets revealed enrichment for angiogenic process networks. A 1.1-kb SF-1-binding region was identified in the angiopoietin 2 (Ang2, ANGPT2) promoter in a highly repetitive region, and SF-1-dependent activation was confirmed in luciferase assays. Angiogenesis is paramount in adrenal development and tumorigenesis, but until now a direct link between SF-1 and vascular remodeling has not been established. We have identified Ang2 as a potentially important novel target of SF-1 in the adrenal gland, indicating that regulation of angiogenesis might be an important additional mechanism by which SF-1 exerts its actions in the adrenal gland.