EMAIL THIS PAGE TO A FRIEND
Optics express
Solution-processed chalcogenide glass for integrated single-mode mid-infrared waveguides.
PMID 21165024
Abstract
Chalcogenide glass materials exhibit a variety of optical properties that make them desirable for near- and mid-infrared communications and sensing applications. However, processing limitations for these photorefractive materials have made the direct integration of waveguides with sources or detectors challenging. Here we demonstrate the viability of two complementary soft lithography methods for patterning and integrating chalcogenide glass waveguides from solution. One method, micro-molding in capillaries (MIMIC), is shown to fabricate multi-mode As(2)S(3) waveguides which are directly integrated with quantum cascade lasers (QCLs). In a second method, we demonstrate the ability of micro-transfer molding (µTM), to produce arrays of single mode rib waveguides (2.5 µm wide and 4.5 µm high) over areas larger than 6 cm(2) while maintaining edge roughness below 5.1 nm. These methods form a suite of processes that can be applied to chalcogenide solutions to create a diverse array of mid-IR optical and photonic structures ranging from <5 to 10's of µm in dimension.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
448060
Arsenic(III) sulfide, ≥99.99%
As2S3