EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Fractional composition of blood serum lipoproteins in mice and rats with Triton WR 1339-induced lipemia.
PMID 21165388
Abstract
We compared fractional composition of blood serum lipoproteins (LP) in female ICR mice and Wistar rats induced by single administration of a nonionic detergent Triton WR 1339 in doses of 300 and 500 mg/kg. Lipemia in animals of both species was characterized by a sharp increase in the concentration of cholesterol and, particularly, of triglycerides in blood serum lipoproteins by the 24th hour after administration of the detergent. We revealed a significant increase in the concentrations of atherogenic VLDL cholesterol (due to VLDL2), intermediate density lipoproteins, and LDL. These changes were more pronounced in rats. The model of lipemia can be used to study the role of fractional composition of lipoproteins and, particularly, of triglycerides in the pathogenesis of atherosclerosis. Moreover, this model holds much promise for evaluation of the efficiency of hypolipidemic drugs (statins and fibrates) in normalizing the increased level of atherogenic cholesterol of VLDL and LDL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8761
Tyloxapol, nonionic surfactant
T0307
Tyloxapol, BioXtra