EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
5-Aza-2'-deoxycytidine increases sialyl Lewis X on MUC1 by stimulating β-galactoside:α2,3-sialyltransferase 6 gene.
PMID 21168523
Abstract
Sialyl Lewis X is a tumor-associated antigen frequently found in the advanced cancers. However, the mechanism for the production of this cancer antigen is not entirely clear. The objective of this study is to examine whether epigenetics is involved in the regulation of the formation of this antigen. We observed an increase of sialyl Lewis X in HCT15 cells, a colon cancer cell line, treated with 5-Aza-2'-deoxycytidine. This treatment enhanced the expression of β-galactoside:α2,3-sialyltransferase 6 gene and sialyl Lewis X on MUC1, and the adherence of these cells to E-selectin under dynamic flow conditions. In addition, 5-Aza-2'-deoxycytidine treatment inhibited methylation of β-galactoside:α2,3-sialyltransferase 6 gene and siRNA knockdown of this gene drastically reduced sialyl Lewis X without affecting MUC1 expression. We conclude that 5-Aza-2'-deoxycytidine treatment increases sialyl Lewis X on MUC1 by stimulating the β-galactoside:α2,3-sialyltransferase 6 gene via inhibition of DNA methylation. Increased sialyl Lewis X by 5-Aza-2'-deoxycytidine raises a concern about the safety of this chemotherapeutic drug. In addition, β-galactoside:α2,3-sialyltransferase 6 gene may be a potential therapeutic target for suppressing tumorigenicity of colon cancer.