EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Biotransformation of quinazoline and phthalazine by Aspergillus niger.
PMID 21169055
Abstract
Cultures of Aspergillus niger NRRL-599 in fluid Sabouraud medium were grown with quinazoline and phthalazine for 7 days. Metabolites were purified by high-performance liquid chromatography and identified by mass spectrometry and proton nuclear magnetic resonance spectroscopy. Quinazoline was oxidized to 4-quinazolinone and 2,4-quinazolinedione, and phthalazine was oxidized to 1-phthalazinone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142026
Benzoyleneurea, 97%
C8H6N2O2
P38706
Phthalazine, 98%
C8H6N2