EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Butein, a tetrahydroxychalcone, suppresses cancer-induced osteoclastogenesis through inhibition of receptor activator of nuclear factor-kappaB ligand signaling.
PMID 21170936
Abstract
Osteoclastogenesis is associated with aging and various age-related inflammatory chronic diseases, including cancer. Receptor activator of nuclear factor-kappaB (NF-κB) ligand (RANKL), a member of the tumor necrosis factor superfamily, has been implicated as a major mediator of bone resorption, suggesting that agents that can suppress RANKL signaling might inhibit osteoclastogenesis, a process closely linked to bone resorption. We therefore investigated whether butein, a tetrahydroxychalcone, could inhibit RANKL signaling and suppress osteoclastogenesis induced by RANKL or tumor cells. We found that human multiple myeloma cells (MM.1S and U266), breast tumor cells (MDA-MB-231) and prostate tumor cells (PC-3) induced differentiation of macrophages to osteoclasts, as indicated by tartrate-resistant acid phosphatase (TRAP)-positive cells, and that butein suppressed this process. The chalcone also suppressed the expression of RANKL by the tumor cells. We further found that butein suppressed RANKL-induced NF-κB activation and that this suppression correlated with the inhibition of IκBα kinase and suppression of phosphorylation and degradation of IκBα, an inhibitor of NF-κB. Finally, butein also suppressed the RANKL-induced differentiation of macrophages to osteoclasts in a dose-dependent and time-dependent manner. Collectively, our results indicate that butein suppresses the osteoclastogenesis induced by tumor cells and by RANKL, by suppression of the NF-κB activation pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B178
Butein, solid
C15H12O5