EMAIL THIS PAGE TO A FRIEND
Organic letters
Photoinduced rearrangements of 3,3'-bis(arylbenzofurans).
PMID 21171574
Abstract
Complex tetracyclic ring systems were assembled by a photoinduced rearrangement of 3,3'-bis(arylbenzofurans). Irradiation of 1 under N(2) atmosphere yielded the benzonaphthofurans 2 in 75-90% yield. When the reaction was conducted under an O(2) atmosphere in the presence of tetracyanoethylene (TCNE) a different reaction pathway was observed leading to the isolation of 3 as the major product. Also, the photochemical properties of these novel structures were investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8809
Tetracyanoethylene, 98%
C6N4