EMAIL THIS PAGE TO A FRIEND
Drugs in R&D
Niacin extended release (ER)/simvastatin (Simcor®): a guide to its use in lipid regulation.
PMID 21171671
Abstract
Oral fixed-dose niacin extended release/simvastatin is associated with clinically relevant improvements in plasma lipid profiles, including lowering of non-high-density lipoprotein cholesterol levels, relative to simvastatin monotherapy in patients with mixed dyslipidemias who had not responded fully to simvastatin monotherapy, and is generally well tolerated.