EMAIL THIS PAGE TO A FRIEND
Scientia pharmaceutica
Synthesis, photochemical and photoinduced antibacterial activity studies of meso-Tetra(pyren-1-yl)porphyrin and its Ni, Cu and Zn complexes.
PMID 21179316
Abstract
The synthesis of the meso-tetra(pyren-1-yl)porphyrin (1) was successfully accomplished by means of the pyrrole condensation with pyrene-1-carb-aldehyde in acidic media. Its metallization was carried out in an almost quantitative yield to obtain the corresponding complexes of Ni(II) (2), Cu(II) (3) and Zn (4). Their photophysical properties such as fluorescence quantum yield and energy transfer to oxygen for an efficient generation of singlet oxygen were determined. Their photophysical and photochemical properties were compared with those of other similar porphyrin derivatives such as tetraphenylporphyrin and tetranaphthylporphyrin. Photochemical studies on their effectiveness as photosensitizer were carried out by means of the photoinduced oxidation of aromatic alcohols like Î-naphthol to naphthoquinone. The antibacterial photoactivity assay for compounds 1â4 was testeted against Escherichia coli (ATCC 8739) and its proliferation and viability were measured by chemiluminescence. An efficient inactivation of E. coli was observed. This was more efficient for compounds 2 and 3, following the direct relationship to high generation of singlet oxygen by these compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144037
1-Pyrenecarboxaldehyde, 99%
C17H10O