EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and biological activity of pyrazolothienotetrahydroisoquinoline and [1,2,4]triazolo[3,4-a]thienotetrahydroisoquinoline derivatives.
PMID 21185109
Abstract
1-hydrazino-3-thioxo-5,6,7,8-tetrahydroisoquinoline-4-carbonitrile (3) was subjected to react with bifunctional compounds namely: acetylacetone, ethyl cyanoacetate, ethyl benzoylactate, diethylmalonate and ethyl acetoacetate to produce pyrazololthienotetrahydroisoquinoline derivatives 6-11. Also, heating of compound (3) with formic acid afforded triazolothienotetrahydroisoquinoline compound 5 which reacted with α-halogenated compounds to afford compounds 13a-e. Compound 13c when heated with triethylorthoformate afforded triazolo derivative 14. Also, compound 6 was used for synthesizing compounds 18-20. Representative compounds of the synthesized triazolo and pyrazolothienotetrahydroisoquinoline products were tested and evaluated as antimicrobial agents.