EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis and antibacterial activity of 3-methylenepyrrolidine formyl hydroxyamino derivatives as novel peptide deformylase inhibitors.
PMID 21185180
Abstract
The synthesis and antibacterial activity of 3-methylenepyrrolidine formyl hydroxyamino derivatives are reported. The antibacterial activities of these derivatives were evaluated to discover SAR at P(1') and P(3') positions, and most of these derivatives exhibit better in vitro antibacterial activity than existing drugs against drug-resistant clinical isolates including MRSA, PRSP, and Haemophilus influenzae.