EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Protective effect of embelin against acetic acid induced ulcerative colitis in rats.
PMID 21185828
Abstract
The aim of the present study is to evaluate the effect of embelin isolated from Embelia ribes on acetic acid induced colitis in rats. Experimental animals received embelin (25 and 50 mg/kg, p.o.) and sulfasalazine (100mg/kg, p.o.) for five consecutive days before induction of colitis by intra-rectal acetic acid (3% v/v) administration and the treatment continued up to 7 days. The colonic mucosal injury was assessed by clinical, macroscopic, biochemical and histopathological examinations. Embelin treatment significantly decreased clinical activity score, gross lesion score, percent affected area and wet colon weight when compared to acetic acid induced controls. The treatment also reduced significantly the colonic myeloperoxidase activity, lipid peroxides and serum lactate dehydrogenase and significantly increased the reduced glutathione. The histopathological studies also confirmed the foregoing findings. The protective effect may be due to its antioxidant and anti-inflammatory activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1406
Embelin, ≥98% (HPLC), powder
C17H26O4