EMAIL THIS PAGE TO A FRIEND
Neuro endocrinology letters
In vivo effect of pinosylvin and pterostilbene in the animal model of adjuvant arthritis.
PMID 21187826
Abstract
The aim of this study was to evaluate the effects of pinosylvin (PIN) and pterostilbene (PTE), natural substances from the stilbenoid group, on the development of adjuvant arthritis in rats. Adjuvant arthritis (AA) was induced by a single intradermal injection of Mycobacterium butyricum in incomplete Freund's adjuvant in male Lewis rats. Our experiments included healthy intact animals as reference controls, arthritic animals without any drug administration, and arthritic animals with administration of PIN and PTE in the oral daily dose of 30 mg/kg b.w. The treatment involved administration of the substances tested from day 0, i.e. the day of immunization, to the experimental day 28. The following parameters were monitored: change of the hind paw volume (HPV) on day 14, 21 and 28, luminol-enhanced chemiluminescence (CL) of the joint and myeloperoxidase (MPO) activity in hind paw joint homogenates (day 28). Arthritic animals treated with PIN showed a decrease in HPV, significantly on days 14 and 28. PIN decreased CL of the joint as well as MPO activity of the joint homogenate, in comparison with untreated animals. PTE had no effect on HPV and MPO activity in hind paw joint homogenates and exerted only a partial effect on luminol-enhanced CL. On the basis of our results we conclude that the effect of PTE on CL was only partial. PIN, on the other hand, had a beneficial anti-inflammatory and antioxidant effect on oxidative stress induced biochemical changes occurring in AA, as determined by all three functional parameters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56297
Pinosylvin, ≥97.0% (HPLC)
C14H12O2