EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Identifying chelators for metalloprotein inhibitors using a fragment-based approach.
PMID 21189019
Abstract
Fragment-based lead design (FBLD) has been used to identify new metal-binding groups for metalloenzyme inhibitors. When screened at 1 mM, a chelator fragment library (CFL-1.1) of 96 compounds produced hit rates ranging from 29% to 43% for five matrix metalloproteases (MMPs), 24% for anthrax lethal factor (LF), 49% for 5-lipoxygenase (5-LO), and 60% for tyrosinase (TY). The ligand efficiencies (LE) of the fragment hits are excellent, in the range of 0.4-0.8 kcal/mol. The MMP enzymes all generally elicit the same chelators as hits from CFL-1.1; however, the chelator fragments that inhibit structurally unrelated metalloenzymes (LF, 5-LO, TY) vary considerably. To develop more advanced hits, one hit from CFL-1.1, 8-hydroxyquinoline, was elaborated at four different positions around the ring system to generate new fragments. 8-Hydroxyquinoline fragments substituted at either the 5- or 7-positions gave potent hits against MMP-2, with IC(50) values in the low micromolar range. The 8-hydroxyquinoline represents a promising new chelator scaffold for the development of MMP inhibitors that was discovered by use of a metalloprotein-focused chelator fragment library.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
232505
1,2-Dihydroxynaphthalene, technical grade
C10H8O2
311308
1,4,7-Triazacyclononane, 95%
C6H15N3
126209
2,3-Dihydroxybenzoic acid, 99%
C7H6O4
122505
2,3-Dihydroxypyridine, 95%
C5H5NO2
P63204
2,3-Pyridinedicarboxylic acid, 99%
C7H5NO4
D109401
2,4-Dihydroxybenzoic acid, 97%
C7H6O4
W379808
2,4-Dihydroxybenzoic acid, ≥97%
C7H6O4
P63603
2,5-Pyridinedicarboxylic acid, 98%
C7H5NO4
D109606
2,6-Dihydroxybenzoic acid, 98%
C7H6O4
02321
2,6-Pyridinedicarboxylic acid, for ion chromatography, ≥99.5% (T)
C7H5NO4
P63808
2,6-Pyridinedicarboxylic acid, 99%
C7H5NO4
412929
2-Ethyl-3-hydroxy-4H-pyran-4-one, 99%
C7H8O3
184055
2-Methoxythiophenol, 97%
C7H8OS
H57602
2-Methyl-8-quinolinol, 98%
C10H9NO
P42800
2-Picolinic acid, ReagentPlus®, 99%
C6H5NO2
T7750
2-Thiouracil, ≥99%
C4H4N2OS
227153
3-Chloro-2,4-pentanedione, 97%
C5H7ClO2
H43407
3-Hydroxy-2-methyl-4-pyrone, 99%
C6H6O3
56197
3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C6H5NO3
152307
3-Hydroxypyridine-2-carboxylic acid, 98%
C6H5NO3
282855
3-Methyl-2,4-pentanedione, mixture of tautomers, technical grade, 85%
C6H10O2
167037
3-Methylsalicylic acid, 97%
C8H8O3
C47000
5-Chloro-8-quinolinol, 95%
C9H6ClNO
152706
5-Fluorosalicylic acid, 97%
C7H5FO3
146161
5-Methylsalicylic acid, 98%
C8H8O3
384305
6-Hydroxypyridine-2-carboxylic acid, 95%
C6H5NO3
462128
6-Methylpyridine-2-carboxylic acid
C7H7NO2
252565
8-Hydroxyquinoline, ACS reagent, 99%
C9H7NO
H6878
8-Hydroxyquinoline, crystalline
C9H7NO
36524
8-Quinolinol, PESTANAL®, analytical standard
C9H7NO