EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Gas chromatography/mass spectrometry determination of mephedrone in drug seizures after derivatization with 2,2,2-trichloroethyl chloroformate.
PMID 21192034