EMAIL THIS PAGE TO A FRIEND
The neurologist
Xylometazoline abuse induced ischemic stroke in a young adult.
PMID 21192193
Abstract
substance abuse is an important cause of ischemic stroke in the young. This includes over-the-counter dietary supplements and cough and cold remedies, which were reported to be an independent risk factor for hemorrhagic stroke. this article describes a young male patient with acute ischemic infarctions in the posterior inferior cerebellar and posterior cerebral artery territories bilaterally, the right cerebral peduncle, the left pontine tegmentum, and lateral pons following abuse of xylometazoline-containing nasal decongestant for 10 years. this is the first report in the literature of posterior circulation strokes because of chronic xylometazoline abuse. We hope to contribute to increase knowledge and awareness of the public about these serious complications of cough-and-cold remedies as well as dietary supplements containing sympathomimetics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X6000
Xylometazoline hydrochloride
C16H24N2 · HCl