EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Furazolidone induced oxidative DNA damage via up-regulating ROS that caused cell cycle arrest in human hepatoma G2 cells.
PMID 21195149
Abstract
Furazolidone (FZD) is an antimicrobial agent that has been shown to have mutagenic, genotoxic and potentially carcinogenic properties when tested in a variety of systems in vitro and in vivo. In this study, we investigated FZD's DNA damaging effect in human hepatoma cells aiming at further defining the molecular mechanism of FZD's cytotoxicity. Addition of FZD resulted in cell growth suppression and cell cycle arrest in S phase accompanied by remarkable DNA strand breaks with increased levels of intracellular reactive oxygen species (ROS) and 8-hydroxydeoxyguanosine. Activities of antioxidases were down-regulated following FZD treatment and antioxidant agent catalase and superoxide dismutase ameliorated FZD's DNA damaging effects. Moreover, FZD caused much more extensive damage to mitochondrial DNA (mtDNA) than to nuclear DNA for which the decrease in mtDNA content correlated with FZD usage in a dose-dependent manner. However, there was no evidence of FZD induced mtDNA mutation in the mitochondrial DNA displacement loop. These results demonstrate that FZD up-regulates the production of intracellular ROS to cause oxidative DNA damage with mtDNA being the most vulnerable targets. Oxidative stress and the injury of mtDNA could be early indicators of FZD-induced cytotoxicity, with the resulting abnormal progression of the cell cycle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9505
Furazolidone
C8H7N3O5
46297
Furazolidone, VETRANAL, analytical standard
C8H7N3O5