EMAIL THIS PAGE TO A FRIEND
Meat science
Effects of sodium lactate and acetic acid derivatives on the quality and sensory characteristics of hot-boned pork sausage patties.
PMID 21195560
Abstract
Sodium lactate and acetic acid derivatives were evaluated for their effects on color retention, microbial growth, and sensory attributes of hot-boned pork sausage patties. Treatments included: (a) sodium lactate (L), (b) buffered vinegar (V), (c) sodium lactate and vinegar mixture (LV), (d) control with BHA/BHT (C), and (e) negative control (NC). Treatments L and LV decreased TPC at day 14 and day 16 when compared to control samples and reduced bacterial numbers up to 18 days. In addition, use of lactate and vinegar increased (P<0.05) acceptability and juiciness and reduced (P<0.05) off-flavor and rancidity when compared to control treatments at day 14. These results revealed that the L and LV sausage patties retained sensory acceptability and shelf-life quality from day 14 through day 17 as opposed to other treatments. Additionally, sausage patties with LV maintained redness and sensory quality throughout 17 days of shelf-life, as compared to other treatments that retained color and quality for 14 days.