EMAIL THIS PAGE TO A FRIEND
Brazilian dental journal
Dentin permeability of the apical third in different groups of teeth.
PMID 21203703
Abstract
This ex vivo study evaluated dentin permeability of the root canal in the apical third of different human groups of teeth. Eighty teeth were used, 8 from each dental group: maxillary and mandibular central incisors, lateral incisors and canines, maxillary first premolars (buccal and palatal roots), mandibular first premolars, and maxillary and mandibular second premolars, totalizing 88 roots that were distributed in 11 groups. The root canals were instrumented, irrigated with 1% NaOCl and 15% EDTA. Roots were immersed in 10% copper sulfate for 30 min and then in 1% rubeanic acid alcohol solution for the same period; this chemical reaction reveals dentin permeability by the formation of copper rubeanate, which is a dark-colored compound. Semi-serial 100-µm-thick cross-sections were obtained from the apical third of the roots. Five sections of each apical third were washed, dehydrated, cleared and mounted on glass slides for examination under optical microscopy. The percentage of copper ion infiltration and the amount of tubular dentin were quantified by morphometric analysis. The penetration of copper ions in the apical third ranged from 4.60 to 16.66%. The mandibular central and lateral incisors presented the highest dentin permeability (16.66%), while the maxillary canines and mandibular second and first premolars presented the lowest dentin permeability (4.60%, 4.80% and 5.71%, respectively; p<0.001). The other teeth presented intermediate permeability. In conclusion, dye penetration into dentin tubules at the apical region is strongly dependent on the group of teeth evaluated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379387
Dithiooxamide, 97%
C2H4N2S2