EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Antimicrobial properties of N3-(iodoacetyl)-L-2,3-diaminopropanoic acid-peptide conjugates.
PMID 2120441
Abstract
Six peptide conjugates consisting of either norvaline, methionine, or lysine and N3-(iodoacetyl)-L-2,3-diaminopropanoic acid--a strong, irreversible inactivator of bacterial and fungal glucosamine-6-phosphate synthase--were synthesized and their antibacterial and antifungal activities were evaluated. Antimicrobial potencies of these peptides were correlated with their transport and cleavage rates inside the cells. Bacteriolysis of Bacillus pumilus cells and inhibition of [14C]glucose incorporation into cell-wall polysaccharides of Candida albicans as a result of glucosamine 6-phosphate inactivation were also observed. Reversal of growth inhibitory effect of these peptides by N-acetylglucosamine in bacteria and fungi suggests the effective delivery of N3-iodoacetyl-L-2,3-diaminopropanoic acid into the cell by a peptide-transport system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15402
Boc-Dap-OH, ≥98.0% (TLC)
C8H16N2O4