EMAIL THIS PAGE TO A FRIEND
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Effects of cetrorelix, a GnRH-receptor antagonist, on gonadal axis in women with functional hypothalamic amenorrhea.
PMID 21204607
Abstract
Gonadotropin Releasing Hormone (GnRH) antagonists (GnRHa) suppress gonadotropin and sex-steroid secretion. In normal women, acute GnRHa administration induces inhibitory effect on pituitary-gonadal axis, followed by Luteinizing Hormone (LH) rebound. Functional hypothalamic amenorrhea (HA) is characterised by impaired gonadotropin secretion and hypogonadism secondary to blunted GnRH pulsatility. We studied the effects of a GnRHa, cetrorelix (CTX 3.0 mg), in six women with HA (age 30.7 ± 3.2 years; BMI 21.5 ± 1.7 kg/m(2)) and six control subjects (CS, 28.2 ± 0.6 years; 22.6 ± 0.9 kg/m(2)) on LH, Follicle-Stimulating Hormone (FSH) and oestradiol levels over 4 h (08.00-12.00 am) before, +24 h and +96 h after CTX; LH, FSH, and oestradiol were also evaluated at +6, +8, +12, +48, +72 h after CTX. CS: CTX reduced (p < 0.05) LH, FSH, and oestradiol (nadir at +12 h, +24 h, and +24 h); LH rebounded at +96 h, FSH and oestradiol recovered at +48 h and +72 h. The 4-h evaluation showed LH and FSH reduction (p < 0.05) at +24 h, with LH rebound at +96 h. HA: CTX reduced (p < 0.05) LH, FSH, and oestradiol, (nadir at +24 h, +48 h, and +48 h, recovery at +48 h, +72 h, and +96 h). The 4-h evaluation showed gonadotropin reduction (p < 0.05) 24 h after CTX, without any rebound effect. One single CTX dose still modulates gonadotropin secretion in HA. Its 'paradoxical' stimulatory effect on gonadotropins needs to be verified after prolonged administration.