EMAIL THIS PAGE TO A FRIEND
Redox report : communications in free radical research
Comments on Vennila and Pugalendi. Vennila L, Pugalendi KV. Protective effect of sesamol against myocardial infarction caused by isoproterenol in Wistar rats. Redox Rep 2010; 15: 36-42.
PMID 21208529
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3003
Sesamol, 98%
C7H6O3