EMAIL THIS PAGE TO A FRIEND
Schweizer Archiv fur Tierheilkunde
[Piglet castration using injection anesthesia: experiences with a combination of azaperone, butorphanol and ketamine].
PMID 21210371
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34223
Azaperone, VETRANAL, analytical standard
C19H22FN3O