EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Photo-allergic contact dermatitis caused by isoamyl p-methoxycinnamate in an 'organic' sunscreen.
PMID 21210828
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44189
Isoamyl 4-methoxycinnamate, analytical standard
C15H20O3