EMAIL THIS PAGE TO A FRIEND
Cell Cycle
?H2A is a component of yeast heterochromatin required for telomere elongation.
PMID 21212735
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-419
Anti-Myc Tag Antibody, clone 9E10, clone 9E10, Upstate®, from mouse