EMAIL THIS PAGE TO A FRIEND
Talanta
Identification of the geometrical isomers of α-linolenic acid using gas chromatography/mass spectrometry with a binary decision tree.
PMID 21215858
Abstract
Gas chromatography, using a highly polar column, low energy (30 eV) electron ionization mass spectrometry and multivariate curve resolution, are combined to obtain the mass spectra of all eight geometrical isomers of α-linolenic acid. A step by step Student's t-test is performed on the m/z 50-294 to identify the m/z by which the geometries of the double bonds could be discriminated. The most intense peak discriminates between cis (m/z 79) and trans (m/z 95) at the central (carbon 12) position. The configuration at carbon 15 is then distinguished by m/z 68 and 236, and finally the geometry at carbon 9 is determined by m/z 93, 173, 191 and 236. A three-question binary tree is developed based on the normalized intensities of these ions by which the identity of any given isomer of α-linolenic is accurately determined. Application of Bayes theorem to data from independent samples shows that the complete configuration is determined correctly with a minimum probability of 87%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6031
Linolenic acid methyl ester isomer mix