EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
The structure of molten AgCl, AgI and their eutectic mixture as studied by molecular dynamics simulations of polarizable ion model potentials.
PMID 21219005
Abstract
The structure of molten AgCl, AgI, and their eutectic mixture Ag(Cl(0.43)I(0.57)) is studied by means of molecular dynamics simulations of polarizable ion model potentials. The corresponding static coherent structure factors reproduce quite well the available neutron scattering data. The qualitative behavior of the simulated partial structure factors and radial distribution functions for molten AgCl and AgI is that predicted by the reverse Monte Carlo modeling of the experimental data. The AgI results are also in qualitative agreement with those calculated from ab initio molecular dynamics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
204404
Silver iodide, 99.999% trace metals basis
AgI
226823
Silver iodide, 99%
AgI